Assay Details
| Gene Symbol: Ptpn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053908.1 | NP_446360.1 | 9 - 10 | 1262 | 81 | |
| GenBank mRNA | AF468653.1 | - | 2 - 3 | 231 | 81 | |
| BC099824.1 | - | 9 - 10 | 1119 | 7932620 | 81 | |
| FQ232747.1 | - | 3 - 4 | 311 | 81 | ||
| U77038.1 | - | 9 - 10 | 1262 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn6
Genomic Map