Assay Details
| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182949.3 | NP_891994.3 | 2 - 3 | 149 | 107 | |
| GenBank mRNA | AB101677.1 | - | 2 - 3 | 149 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss27
Genomic Map