Assay Details
| Gene Symbol: Rimklb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237315.2 | XP_006237377.1 | 5 - 6 | 2918 | 71 | |
| XM_006237316.2 | XP_006237378.1 | 5 - 6 | 875 | 71 | ||
| XM_006237317.2 | XP_006237379.1 | 5 - 6 | 912 | 71 | ||
| XM_008763276.1 | XP_008761498.1 | 6 - 7 | 1100 | 71 | ||
| XM_342749.7 | XP_342750.3 | 5 - 6 | 833 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rimklb
Genomic Map