Assay Details
| Gene Symbol: Ceacam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169579.1 | NP_001163050.1 | 4 - 5 | 1073 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam16| Gene Symbol: Ceacam16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169579.1 | NP_001163050.1 | 4 - 5 | 1073 | 71 | |
Set Membership: |
|