Assay Details
| Gene Symbol: Polr2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282325.1 | NP_001269254.1 | 1 - 2 | 83 | 87 | |
| GenBank mRNA | AW144706.1 | - | 1 - 2 | 83 | 87 | |
| EX488432.1 | - | 1 - 2 | 54 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001282325.1 transcripts.
This assay is in the 5'UTR region of the NM_001282325.1 transcripts.
Set Membership: |
|

Polr2k
Genomic Map