Assay Details
| Gene Symbol: Spint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004265.2 | NP_001004265.2 | 1 - 2 | 689 | 64 | |
| XM_006234763.1 | XP_006234825.1 | 2 - 3 | 701 | 64 | ||
| XM_006234764.2 | XP_006234826.1 | 2 - 3 | 627 | 64 | ||
| XM_008762100.1 | XP_008760322.1 | 2 - 3 | 701 | 64 | ||
| GenBank mRNA | AB154834.1 | - | 1 - 2 | 462 | 64 | |
| BC097253.1 | - | 1 - 2 | 689 | 7459422 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spint1
Genomic Map