Assay Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047918.2 | NP_001041383.2 | 13 - 14 | 2208 | 59 | |
| GenBank mRNA | FQ231577.1 | - | 12 - 13 | 1888 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bank1
Genomic Map