Assay Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134465.2 | NP_604460.2 | 5 - 6 | 756 | 117 | |
| XM_008769915.1 | XP_008768137.1 | 5 - 6 | 714 | 117 | ||
| XM_008769916.1 | XP_008768138.1 | 5 - 6 | 739 | 117 | ||
| GenBank mRNA | AF404510.1 | - | 5 - 6 | 699 | 117 | |
| BC078740.1 | - | 5 - 6 | 756 | 7108137 | 117 | |
| FQ232217.1 | - | 5 - 6 | 742 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crlf2
Genomic Map