Assay Details
| Gene Symbol: Polr3gl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109571.1 | NP_001103041.1 | 5 - 6 | 498 | 86 | |
| XM_008761331.1 | XP_008759553.1 | 5 - 6 | 581 | 86 | ||
| GenBank mRNA | BC168229.1 | - | 5 - 6 | 457 | 7377062 | 86 |
| FQ214367.1 | - | 5 - 6 | 484 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr3gl
Genomic Map