Assay Details
| Gene Symbol: Ndufb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106360.2 | NP_001099830.1 | 2 - 3 | 218 | 92 | |
| GenBank mRNA | BC166889.1 | - | 2 - 3 | 218 | 6891455 | 92 |
| BC166905.1 | - | 2 - 3 | 218 | 6887475 | 92 | |
| BC166906.1 | - | 2 - 3 | 218 | 6888064 | 92 | |
| FQ217125.1 | - | 2 - 3 | 212 | 92 | ||
| FQ217646.1 | - | 2 - 3 | 221 | 92 | ||
| FQ223847.1 | - | 2 - 3 | 233 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufb8
Genomic Map