Assay Details
| Gene Symbol: Rpl7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_228615.8 | XP_228615.3 | 1 - 2 | 327 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_228615.8 transcripts.
Set Membership: |
|

Rpl7l1
Genomic Map