Assay Details
| Gene Symbol: Tmprss7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105882.2 | NP_001099352.2 | 17 - 18 | 2319 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss7
Genomic Map