Assay Details
| Gene Symbol: Tmprss11f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770104.1 | XP_008768326.1 | 8 - 9 | 1073 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11f| Gene Symbol: Tmprss11f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770104.1 | XP_008768326.1 | 8 - 9 | 1073 | 67 | |
Set Membership: |
|