Assay Details
| Gene Symbol: Clcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207615.2 | NP_997498.2 | 2 | 593 | 168 | |
| GenBank mRNA | AY365006.1 | - | 2 | 340 | 168 | |
| BC098643.1 | - | 2 | 593 | 7123848 | 168 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clcf1
Genomic Map