Assay Details
| Gene Symbol: Trex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024989.1 | NP_001020160.1 | 2 | 856 | 152 | |
| GenBank mRNA | BC091242.1 | - | 2 | 856 | 152 | |
| FQ233170.1 | - | 2 | 906 | 152 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trex1
Genomic Map