Assay Details
| Gene Symbol: Ceacam6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045097.1 | 6 - 7 | 1400 | 86 | ||
| GenBank mRNA | AB434131.1 | - | 5 - 6 | 1228 | 86 | |
| BC078962.1 | - | 6 - 7 | 1400 | 7111028 | 86 | |
| Gene Symbol: LOC103689938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759078.1 | XP_008757300.1 | 1510 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam6
Genomic Map