Assay Details
| Gene Symbol: Ceacam6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045097.1 | 7 - 8 | 1518 | 83 | ||
| GenBank mRNA | BC078962.1 | - | 7 - 8 | 1518 | 7111028 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ceacam6
Genomic Map