Assay Details
| Gene Symbol: Prss36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011560.2 | NP_001011560.2 | 6 - 7 | 722 | 71 | |
| XM_008759913.1 | XP_008758135.1 | 6 - 7 | 1576 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prss36
Genomic Map