Assay Details
| Gene Symbol: Trex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024989.1 | NP_001020160.1 | 2 | 918 | 91 | |
| GenBank mRNA | BC091242.1 | - | 2 | 918 | 91 | |
| FQ233170.1 | - | 2 | 968 | 91 | ||
More Information
Set Membership: |
|

Trex1
Genomic Map