Assay Details
| Gene Symbol: SOCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123196.1 | NP_001116668.1 | 1 | 456 | 112 | |
| XM_021066253.1 | XP_020921912.1 | 787 | 112 | |||
| GenBank mRNA | AY785556.1 | - | 1 | 432 | 112 | |
| DQ644577.1 | - | 1 | 465 | 112 | ||
| HM045422.1 | - | 1 | 456 | 112 | ||
More Information
Set Membership: |
|

SOCS3
Genomic Map