Assay Details
| Gene Symbol: RPL7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123192.1 | NP_001116664.1 | 3 - 4 | 592 | 71 | |
| GenBank mRNA | AK237590.1 | - | 3 - 4 | 303 | 71 | |
| AK391110.1 | - | 3 - 4 | 332 | 71 | ||
| AY609649.2 | - | 3 - 4 | 565 | 71 | ||
More Information
Set Membership: |
|

RPL7L1
Genomic Map