Assay Details
| Gene Symbol: NECTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001262.1 | NP_001001262.1 | 2 - 3 | 426 | 62 | |
| XM_021102215.1 | XP_020957874.1 | 1365 | 62 | |||
| GenBank mRNA | AF308632.1 | - | 2 - 3 | 426 | 62 | |
More Information
Set Membership: |
|

NECTIN1
Genomic Map