Assay Details
| Gene Symbol: POLR2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243341.1 | NP_001230270.1 | 4 - 5 | 461 | 73 | |
| XM_005670048.3 | XP_005670105.1 | 1526 | 73 | |||
| XM_021068495.1 | XP_020924154.1 | 1756 | 73 | |||
| GenBank mRNA | AK235943.1 | - | 4 - 5 | 480 | 73 | |
| BP463051.1 | - | 4 - 5 | 463 | 73 | ||
| BW980083.1 | - | 4 - 5 | 422 | 73 | ||
More Information
Set Membership: |
|

POLR2H
Genomic Map