Assay Details
| Gene Symbol: PPP1R7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243902.1 | NP_001230831.1 | 6 - 7 | 629 | 62 | |
| XM_005654201.3 | XP_005654258.1 | 503 | 62 | |||
| XM_013982499.2 | XP_013837953.1 | 689 | 62 | |||
| GenBank mRNA | AK235212.1 | - | 6 - 7 | 629 | 62 | |
| AK398801.1 | - | 6 - 7 | 611 | 62 | ||
More Information
Set Membership: |
|

PPP1R7
Genomic Map