Assay Details
| Gene Symbol: SPINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244422.2 | NP_001231351.1 | 9 - 10 | 1637 | 59 | |
| XM_005659734.3 | XP_005659791.1 | 1585 | 59 | |||
| XM_013993158.2 | XP_013848612.1 | 1537 | 59 | |||
| XM_013993159.2 | XP_013848613.1 | 1585 | 59 | |||
| XM_021101376.1 | XP_020957035.1 | 1754 | 59 | |||
| XM_021101449.1 | XP_020957108.1 | 1705 | 59 | |||
| GenBank mRNA | AK240186.1 | - | 9 - 10 | 1642 | 59 | |
More Information
Set Membership: |
|

SPINT1
Genomic Map