Assay Details
| Gene Symbol: AP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244452.1 | NP_001231381.1 | 4 - 5 | 484 | 63 | |
| XM_021093322.1 | XP_020948981.1 | 548 | 63 | |||
| XM_021093323.1 | XP_020948982.1 | 548 | 63 | |||
| GenBank mRNA | AK234254.1 | - | 4 - 5 | 422 | 63 | |
| AK392084.1 | - | 4 - 5 | 418 | 63 | ||
| CJ014998.1 | - | 4 - 5 | 423 | 63 | ||
| FS667614.1 | - | 4 - 5 | 418 | 63 | ||
More Information
Set Membership: |
|

AP2S1
Genomic Map