Assay Details
| Gene Symbol: LOC100513671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101397.1 | XP_020957056.1 | 449 | 127 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100513671| Gene Symbol: LOC100513671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101397.1 | XP_020957056.1 | 449 | 127 | ||
Set Membership: |
|