Assay Details
Gene Symbol: KIAA0513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005664311.3 | XP_005664368.1 | 3460 | 120 | ||
XM_013998146.2 | XP_013853600.1 | 3660 | 120 | |||
XM_021093717.1 | XP_020949376.1 | 6941 | 120 | |||
XM_021093718.1 | XP_020949377.1 | 6668 | 120 | |||
XM_021093719.1 | XP_020949378.1 | 3932 | 120 | |||
XM_021093720.1 | XP_020949379.1 | 3363 | 120 | |||
XM_021093721.1 | XP_020949380.1 | 3450 | 120 | |||
GenBank mRNA | AK235824.1 | - | 1 | 1221 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
