Assay Details
| Gene Symbol: MRPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131531.4 | XP_003131579.1 | 586 | 71 | ||
| XM_005653946.3 | XP_005654003.1 | 666 | 71 | |||
| GenBank mRNA | AK348091.1 | - | 4 - 5 | 545 | 71 | |
| AK394533.1 | - | 4 - 5 | 552 | 71 | ||
More Information
Set Membership: |
|

MRPL10
Genomic Map