Assay Details
| Gene Symbol: TREX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070628.1 | XP_020926287.1 | 1055 | 100 | ||
| XM_021070629.1 | XP_020926288.1 | 2727 | 100 | |||
| GenBank mRNA | AK234684.1 | - | 2 | 1024 | 100 | |
| AK398273.1 | - | 2 | 1039 | 100 | ||
More Information
Set Membership: |
|

TREX1
Genomic Map