Assay Details
| Gene Symbol: polr3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203988.1 | NP_989319.1 | 2 - 3 | 336 | 65 | |
| GenBank mRNA | BC064209.1 | - | 2 - 3 | 336 | 65 | |
More Information
Set Membership: |
|

polr3f
Genomic Map