Assay Details
| Gene Symbol: ppp2r1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204074.1 | NP_989405.1 | 4 - 5 | 513 | 66 | |
| GenBank mRNA | BC064863.1 | - | 4 - 5 | 513 | 66 | |
More Information
Set Membership: |
|

ppp2r1a
Genomic Map