Assay Details
| Gene Symbol: ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006730.1 | NP_001006731.1 | 7 - 8 | 672 | 70 | |
| GenBank mRNA | BC075482.1 | - | 7 - 8 | 672 | 70 | |
More Information
Set Membership: |
|

ppp1r7
Genomic Map