Assay Details
| Gene Symbol: pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006774.1 | NP_001006775.1 | 1 - 2 | 140 | 70 | |
| GenBank mRNA | BC075576.1 | - | 1 - 2 | 140 | 70 | |
| CR855580.2 | - | 1 - 2 | 117 | 70 | ||
More Information
Set Membership: |
|

pdgfb
Genomic Map