Assay Details
| Gene Symbol: rpl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011414.1 | NP_001011414.1 | 2 - 3 | 127 | 73 | |
| GenBank mRNA | BC087976.1 | - | 2 - 3 | 127 | 73 | |
| CU025172.1 | - | 2 - 3 | 119 | 73 | ||
| CU464101.1 | - | 1281 | 73 | |||
| Gene Symbol: LOC100493057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002936246.4 | XP_002936292.1 | 2 - 3 | 209 | 73 | |
| GenBank mRNA | BC077707.1 | - | 2 - 3 | 148 | 73 | |
More Information
Set Membership: |
|

rpl32
Genomic Map