Assay Details
| Gene Symbol: mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017222.2 | NP_001017222.1 | 4 - 5 | 374 | 61 | |
| XM_012953002.2 | XP_012808456.1 | 4 - 5 | 61 | |||
| GenBank mRNA | CR760332.2 | - | 4 - 5 | 374 | 61 | |
More Information
Set Membership: |
|

mrpl4
Genomic Map