Assay Details
| Gene Symbol: clcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016067.2 | NP_001016067.1 | 2 - 3 | 224 | 73 | |
| GenBank mRNA | BC118886.1 | - | 2 - 3 | 202 | 73 | |
| CR762259.2 | - | 2 - 3 | 224 | 73 | ||
More Information
Set Membership: |
|

clcf1
Genomic Map