Assay Details
| Gene Symbol: cplane2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016685.2 | NP_001016685.1 | 3 - 4 | 496 | 55 | |
| GenBank mRNA | BC170963.1 | - | 3 - 4 | 445 | 55 | |
| CR760197.2 | - | 3 - 4 | 496 | 55 | ||
More Information
Set Membership: |
|

cplane2
Genomic Map