Assay Details
| Gene Symbol: prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016980.2 | NP_001016980.1 | 5 - 6 | 682 | 109 | |
| GenBank mRNA | CR848351.2 | - | 5 - 6 | 682 | 109 | |
More Information
Set Membership: |
|

prss8
Genomic Map