Assay Details
| Gene Symbol: ras-dva2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044409.1 | NP_001037874.1 | 1 | 456 | 65 | |
| GenBank mRNA | DQ278180.1 | - | 1 | 456 | 65 | |
More Information
Set Membership: |
|

ras-dva2
Genomic Map