Assay Details
| Gene Symbol: polr3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078929.1 | NP_001072397.1 | 4 - 5 | 306 | 73 | |
| XM_012961319.2 | XP_012816773.1 | 4 - 5 | 453 | 73 | ||
| XM_012961320.2 | XP_012816774.1 | 4 - 5 | 432 | 73 | ||
| XM_012961321.2 | XP_012816775.1 | 4 - 5 | 427 | 73 | ||
| GenBank mRNA | BC121597.1 | - | 4 - 5 | 306 | 73 | |
More Information
Set Membership: |
|

polr3h
Genomic Map