Assay Details
| Gene Symbol: ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126936.1 | NP_001120408.1 | 1404 | 109 | ||
| GenBank mRNA | BC161124.1 | - | 1404 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ndor1| Gene Symbol: ndor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126936.1 | NP_001120408.1 | 1404 | 109 | ||
| GenBank mRNA | BC161124.1 | - | 1404 | 109 | ||
Set Membership: |
|