Assay Details
| Gene Symbol: cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128638.1 | NP_001122110.1 | 15 - 16 | 2568 | 66 | |
| XM_012966667.2 | XP_012822121.1 | 14 - 15 | 2813 | 66 | ||
| GenBank mRNA | BC093450.1 | - | 15 - 16 | 2505 | 66 | |
| BC121224.1 | - | 15 - 16 | 2506 | 66 | ||
| BC167455.1 | - | 15 - 16 | 2568 | 66 | ||
| CT030464.1 | - | 11 - 12 | 1692 | 66 | ||
More Information
Set Membership: |
|

cbl
Genomic Map