Assay Details
| Gene Symbol: LOC100285011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157906.1 | NP_001151378.1 | 278 | 91 | ||
| GenBank mRNA | EU970500.1 | - | 278 | 91 | ||
More Information
Set Membership: |
|

LOC100285011
Genomic Map