accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-13-0

IUPAC Name: 1-ethenyl-4-nitrobenzene
Synonyms: 1-ethenyl-4-nitrobenzene
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: YFZHODLXYNDBSM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)