Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrostyrene, 98%, Thermo Scientific™

Catalog No. AAH3011814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3011814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3011814 Supplier Thermo Scientific Chemicals Supplier No. H3011814

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Can be used to introduce amino groups into polymers. The thermal degradation of styrene-4-nitrostyrene copolymers (SNS) has been studied using differential thermal analysis (DTA) and thermogravimetry (TGA) under isothermal and dynamic conditions in dynamic nitrogen. 4—Nitrostyrene gives isomeric (4-nitropheny1)-propiona1dehydes in a 96% yield.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air, heat. Store at 4 deg C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-13-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00041254
InChI Key YFZHODLXYNDBSM-UHFFFAOYSA-N
Synonym 4-nitrostyrene, 1-nitro-4-vinylbenzene, p-nitrostyrene, benzene, 1-ethenyl-4-nitro, ccris 1674, styrene, p-nitro, benzene, 1-ethenyl-4-nitro-9ci, nitrovinylbenzene, 4-nitro-styrene, p-nitrophenylethylene
PubChem CID 7481
IUPAC Name 1-ethenyl-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(C=C)C=C1

Specifications

Melting Point 25°C to 28°C
Density 1.163
Boiling Point 93°C to 96°C (3 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.6045
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Nitrostyrene
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H341
Hazard Statement GHS H Statement
H341-H302-H312-H332-H315-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
RTECSNumber WL5470000
TSCA No
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.