accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO(E)

CAS RN 100-31-2

IUPAC Name: 4-[(1E)-2-(4-carboxyphenyl)ethenyl]benzoic acid
Synonyms: 4-[(1E)-2-(4-carboxyphenyl)ethenyl]benzoic acid
Molecular Weight (g/mol): 268.27
Molecular Formula: C16H12O4
InChi Key: SBBQDUFLZGOASY-OWOJBTEDSA-N
SMILES: OC(=O)C1=CC=C(\C=C\C2=CC=C(C=C2)C(O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)