Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Stilbenedicarboxylic Acid 95.0+%, TCI America™
SDP

Catalog No. S055725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S055725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S055725G Supplier TCI America Supplier No. S055725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-31-2
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00013994
InChI Key SBBQDUFLZGOASY-OWOJBTEDSA-N
Synonym 4,4'-stilbenedicarboxylic acid, stilbene-4,4'-dicarboxylic acid, 4-e-2-4-carboxyphenyl ethenyl benzoic acid, 4,4'-ethene-1,2-diyl dibenzoic acid, 4,4'-stilbenedicarboxylicacid, z-4,4'-ethene-1,2-diyl dibenzoic acid, 4-1e-2-4-carboxyphenyl vinyl benzoic acid, 4,4'-ethene-1,2-diyldibenzoicacid, cis-stilbene-4,4'-dicarboxylic acid, 4,4'-dicarboxystilbene
PubChem CID 5374688
IUPAC Name 4-[(E)-2-(4-carboxyphenyl)ethenyl]benzoic acid
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Quantity 25 g
Formula Weight 268.27
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Stilbenedicarboxylic Acid
EINECSNumber (4)-0292
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.