accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-37-8

IUPAC Name: 2-(diethylamino)ethan-1-ol
Synonyms: DEAE N,N-diethyl-2-aminoethanol β-hydroxytriethylamine deae-sepharose diethyl ethanolamine
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: BFSVOASYOCHEOV-UHFFFAOYSA-N
SMILES: CCN(CC)CCO
Molecular Weight (g/mol) 
  • (10)
  • (3)
Boiling Point 
  • (4)
  • (3)
  • (5)
Percent Purity 
  • (4)
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (5)
  • (1)
  • (3)
  • (10)
  • (3)
  • (4)
  • (3)
  • (5)
  • (4)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (5)
  • (1)