Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diethylaminoethanol, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600111 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-111 200 L Amber Glass Bottle
18-600-107 100 mL Amber Glass Bottle
18-600-109 500 mL Amber Glass Bottle
18-600-110 4 L Amber Glass Bottle
18-600-108 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-111 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1016200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D1016, 100-37-8, C6H15NO

Spectrum™ Chemical 2-Diethylaminoethanol, Reagent is used as a precursor chemical to procaine. In condensate and steam lines, it is used as a corrosion inhibitor because it neutralizes carbonic acid and scavenging oxygen. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 100-37-8
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
IUPAC Name 2-(diethylamino)ethan-1-ol
SMILES CCN(CC)CCO

Specifications

CAS Max % 100%
Boiling Point 163°C
Packaging Amber Glass Bottle
Quantity 200 L
UN Number UN2686
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-Diethylaminoethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.